Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoxetine Related Compound B, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022991 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2991 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2991 Supplier MilliporeSigma Supelco Supplier No. PHR20415ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23580-89-4
Flash Point Not Applicable
Molecular Formula C10H15N
Linear Formula C10H15N
Quantity 5 mL
Synonym N-Methyl-N-(3-phenylpropyl)amine
Molecular Weight (g/mol) 149.23
Formula Weight 149.23
Grade Certified Reference Material
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.