Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoxetine Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018051 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8051 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8051 Supplier MilliporeSigma Supelco Supplier No. PHR195330MG
Add to Cart
Add to Cart

Specifications

CAS 1026723-45-4
Linear Formula C21H22F3 NO4
MDL Number MFCD18382332
Synonym N-Methyl-N-[3-phenyl-3-(4-trifluoromethyl-phenoxy)-propyl]-succinamic acid
Type Certified Reference Material
Quantity 30 mg
Formula Weight 409.40
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.