Learn More
Selleck Chemical LLC Fluoxetine HCl 25mg 56296-78-7 Lilly110140
SDP
Supplier:  Selleck Chemical LLC S133325MG
Encompass_Preferred
Fluoxetine HCl (Lilly110140) is a selective serotonin-reuptake inhibitor (SSRI) at the neuronal membrane, used in the treatment of depression. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-627-2