Learn More
TARGETMOL CHEMICALS INC FLUPAROXAN HYDROCHLORIDE 5MG
SDP
Catalog No. 502579134
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Fluparoxan hydrochloride is a highly selective and potent (alpha)2-adrenoceptor antagonist for the prevention amelioration or treatment of neurodevelopmental disorders and neurodegenerative diseases. purity: 99%
Catalog No. 50-257-9134 Supplier TARGETMOL CHEMICALS INC Supplier No. T775655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Fluparoxan hydrochloride is a highly selective and potent (alpha)2-adrenoceptor antagonist for the prevention amelioration or treatment of neurodevelopmental disorders and neurodegenerative diseases. purity: 99%