Learn More
Medchemexpress LLC HY-15856B 5mg , Flupentixol dihydrochloride CAS:2413-38-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15856B5MG
Encompass_Preferred
HY-15856B 5mg Flupentixol dihydrochloride CAS: 2413-38-9 Flupentixol is a high potency thioxanthene with D1 and D2 dopamine receptor antagonism. Flupentixol is used in therapy of schizophrenia as well as in anxiolytic and depressive disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1488