Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flupentixol dihydrochloride | 2413-38-9 | 98.5% | 507.44 | C23H27Cl2F3N2OS | 10 MG
SDP

Catalog No. 5000287970
Encompass_Preferred

Flupentixol dihydrochloride is the dihydrochloride salt of flupentixol, provided as a research-grade solid for laboratory and analytical applications. Typical purity is 98.52%, with batch-specific certificates reported up to 99.73%. The compound functions as a dopamine D1/D2 receptor antagonist and has reported PI3Kα inhibitory activity.

  • Chemical form: dihydrochloride salt, solid.
  • Pharmacology: D1/D2 dopamine receptor antagonist and reported PI3Kα inhibitor.
  • Purity: typically 98.5%; batch-specific purity may be higher.
  • Appearance: solid.
  • Chemical identifiers: CAS 2413-38-9; molecular weight 507.44; formula C23H27Cl2F3N2OS.
  • Storage: store sealed at 4°C, protect from moisture and light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended use: research and analytical applications, not for human or therapeutic use.

Catalog No. 50-002-87970 Supplier Medchemexpress LLC Supplier No. HY15856B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flupentixol dihydrochloride is the dihydrochloride salt of flupentixol, provided as a research-grade solid for laboratory and analytical applications. Typical purity is 98.52%, with batch-specific certificates reported up to 99.73%. The compound functions as a dopamine D1/D2 receptor antagonist and has reported PI3Kα inhibitory activity.

  • Chemical form: dihydrochloride salt, solid.
  • Pharmacology: D1/D2 dopamine receptor antagonist and reported PI3Kα inhibitor.
  • Purity: typically 98.5%; batch-specific purity may be higher.
  • Appearance: solid.
  • Chemical identifiers: CAS 2413-38-9; molecular weight 507.44; formula C23H27Cl2F3N2OS.
  • Storage: store sealed at 4°C, protect from moisture and light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended use: research and analytical applications, not for human or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.