Learn More
Cayman Chemical FluphenazIn decnoate 10mg
SDP
Catalog No. 502942077
Encompass_Preferred
A prodrug form of fluphenazine; induces random mouth movements in rats; increases serum levels of prolactin and decreases LH and FSH, as well as decreases desire to mate and litter size, in male rats at 3 mg/kg per day; It increases lethargy and passive contact and reduces self-grooming, social grooming, auditory startle response, and propensity to attack other animals in monkeys
Catalog No. 50-294-2077 Supplier Cayman Chemical Supplier No. 3990310 MG
A prodrug form of fluphenazine; induces random mouth movements in rats; increases serum levels of prolactin and decreases LH and FSH, as well as decreases desire to mate and litter size, in male rats at 3 mg/kg per day; It increases lethargy and passive contact and reduces self-grooming, social grooming, auditory startle response, and propensity to attack other animals in monkeys