Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluphenazine, 98+%, Thermo Scientific™

Catalog No. AAJ6117903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6117903 1 g
AAJ6117906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6117903 Supplier Thermo Scientific Chemicals Supplier No. J6117903

Dopamine D1 receptor antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-23-8
Molecular Formula C22H26F3N3OS
Molecular Weight (g/mol) 437.525
MDL Number MFCD00242759
InChI Key PLDUPXSUYLZYBN-UHFFFAOYSA-N
Synonym fluphenazine, triflumethazine, fluorophenazine, fluorphenazine, fluorfenazine, siqualon, phthorphenazine, ftorphenazine, siqualine, vespazine
PubChem CID 3372
ChEBI CHEBI:5123
IUPAC Name 2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethanol
SMILES C1CN(CCN1CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F)CCO

Specifications

Melting Point 268°C to 274°C
Color Brown to Yellow
Quantity 1 g
UN Number UN2811
Merck Index 14,4189
Formula Weight 437.52
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Fluphenazine
Hazard Category Danger
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber TL9730000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.