Learn More
Medchemexpress LLC Fluphenazine-d8 dihydrochloride | 1323633-98-2 | 99.8% | 518.49 g·mol^-1 | C22H20D8Cl2F3N3OS | 10 MG
SDP
Catalog No. 5000299139
Encompass_Preferred
Fluphenazine-d8 dihydrochloride is a deuterium-labeled form of fluphenazine dihydrochloride supplied as a solid research reagent for use as an isotopic internal standard or tracer in biochemical, pharmacokinetic, and metabolic studies. It is intended for research use only and is supplied in a small pack for analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry and metabolic studies.
  • High chemical purity suitable for quantitative analysis (reported 99.8%).
  • Supplied as a solid, stable isotope reagent for laboratory research.
  • Compact pack size for small-scale experiments and method development.
Catalog No. 50-002-99139 Supplier Medchemexpress LLC Supplier No. HYA0081S10MG
Fluphenazine-d8 dihydrochloride is a deuterium-labeled form of fluphenazine dihydrochloride supplied as a solid research reagent for use as an isotopic internal standard or tracer in biochemical, pharmacokinetic, and metabolic studies. It is intended for research use only and is supplied in a small pack for analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry and metabolic studies.
  • High chemical purity suitable for quantitative analysis (reported 99.8%).
  • Supplied as a solid, stable isotope reagent for laboratory research.
  • Compact pack size for small-scale experiments and method development.