Learn More
Medchemexpress LLC Fluphenazine-d8 dihydrochloride | 99.3% | C22H20D8Cl2F3N3OS | 5 MG
SDP
Catalog No. 5000432172
Encompass_Preferred
Fluphenazine-d8 (dihydrochloride) is the deuterium labeled Fluphenazine dihydrochloride. It is a phenothiazine-class D1DR and D2DR inhibitor; used to deliver Fluphenazine to biological systems in studies probing the effects and metabolic fates of this commonly used dopamine antagonist.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32172 Supplier Medchemexpress LLC Supplier No. HYA0081S5MG
Fluphenazine-d8 (dihydrochloride) is the deuterium labeled Fluphenazine dihydrochloride. It is a phenothiazine-class D1DR and D2DR inhibitor; used to deliver Fluphenazine to biological systems in studies probing the effects and metabolic fates of this commonly used dopamine antagonist.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.