Learn More
TARGETMOL CHEMICALS INC Fluphenazine dihydrochloride
SDP
Catalog No. 502880530
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Fluphenazine dihydrochloride (Prolixin) is an inhibitor of phenothiazine-class D1DR and D2DR; In studies, Fluphenazine can be used to probe the effects and metabolic process of this commonly used dopamine antagonist. Purity 100%
Catalog No. 50-288-0530 Supplier TARGETMOL CHEMICALS INC Supplier No. T0068500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Fluphenazine dihydrochloride (Prolixin) is an inhibitor of phenothiazine-class D1DR and D2DR; In studies, Fluphenazine can be used to probe the effects and metabolic process of this commonly used dopamine antagonist. Purity 100%