Learn More
Selleck Chemical LLC Fluphenazine dihydrochloride S4569-500mg
SDP
Catalog No. 503110436
Encompass_Preferred
Fluphenazine dihydrochloride (Prolixin) is a phenothiazine-class D1DR and D2DR inhibitor It also inhibits Histamine H1 Receptor
Catalog No. 50-311-0436 Supplier Selleck Chemical LLC Supplier No. S4569500MG
Add to Cart
Add to Cart
Fluphenazine dihydrochloride (Prolixin) is a phenothiazine-class D1DR and D2DR inhibitor It also inhibits Histamine H1 Receptor