Learn More
Cayman Chemical Fluphenazine (hydrochloride), Purity: ≥95%, CAS: 146-56-5, 1 g
SDP
Catalog No. 502964449
Encompass_Preferred
A D1 and D2 receptor antagonist (Kis = 5 and 0.63 nM, respectively); binds to histamine H1, serotonin 5-HT2A, and 5-HT2B receptors (Kis = 45, 17, and 82 nM, respectively); acts as an inverse agonist at H1 and 5-HT2A receptors; a typical antipsychotic compound that induces extrapyramidal symptoms in monkeys
Catalog No. 50-296-4449 Supplier Cayman Chemical Supplier No. 235551 G
A D1 and D2 receptor antagonist (Kis = 5 and 0.63 nM, respectively); binds to histamine H1, serotonin 5-HT2A, and 5-HT2B receptors (Kis = 45, 17, and 82 nM, respectively); acts as an inverse agonist at H1 and 5-HT2A receptors; a typical antipsychotic compound that induces extrapyramidal symptoms in monkeys