Learn More
Cayman Chemical Fluphenazine-N-2-chloroethane (hydrochloride), Purity: ≥95%, CAS: 3892-78-2, 100 mg
SDP
Catalog No. 502968397
Encompass_Preferred
A derivative of fluphenazine that contains an alkylating chlorethylamine chain, which produces irreversible protein binding; a relatively selective, irreversible antagonist of D2 receptors both in vitro (IC50 = 100 nM) and in vivo; irreversibly inhibits calmodulin at higher doses (IC50 = 10 µM)
Catalog No. 50-296-8397 Supplier Cayman Chemical Supplier No. 18427100 MG
A derivative of fluphenazine that contains an alkylating chlorethylamine chain, which produces irreversible protein binding; a relatively selective, irreversible antagonist of D2 receptors both in vitro (IC50 = 100 nM) and in vivo; irreversibly inhibits calmodulin at higher doses (IC50 = 10 µM)