Learn More
Cayman Chemical FluphenazInN2chLRoethn 10mg
SDP
Catalog No. 502967968
Encompass_Preferred
A derivative of fluphenazine that contains an alkylating chlorethylamine chain, which produces irreversible protein binding; a relatively selective, irreversible antagonist of D2 receptors both in vitro (IC50 = 100 nM) and in vivo; irreversibly inhibits calmodulin at higher doses (IC50 = 10 µM)
Catalog No. 50-296-7968 Supplier Cayman Chemical Supplier No. 1842710 MG
A derivative of fluphenazine that contains an alkylating chlorethylamine chain, which produces irreversible protein binding; a relatively selective, irreversible antagonist of D2 receptors both in vitro (IC50 = 100 nM) and in vivo; irreversibly inhibits calmodulin at higher doses (IC50 = 10 µM)