Learn More
Cayman Chemical Flupirtine-d4 (hydrochloride), Purity: ≥99% deuterated forms (d1-d4), CAS: 1324717-75-0, 1 mg
SDP
Catalog No. 502939503
Encompass_Preferred
An internal standard for the quantification of flupirtine by GC- or LC-MS
Catalog No. 50-293-9503 Supplier Cayman Chemical Supplier No. 286951 MG
An internal standard for the quantification of flupirtine by GC- or LC-MS