Learn More
Medchemexpress LLC Flupirtine Maleate | 75507-68-5 | 100.0% | 420.39 | 100 MG
SDP
Catalog No. 5000390676
Encompass_Preferred
Flupirtine Maleate is an orally active, blood-brain barrier-crossing non-opioid analgesic and neuroprotective agent. It functions as a neuronal potassium channel opener (Kv7 activator), an NMDA receptor antagonist, and a GABA receptor activator. This agent is suitable for research related to focal cerebral ischemia, pain, Alzheimer's disease, or multiple sclerosis.
  • Stabilizes blood-brain barrier integrity.
  • Reduces oxidative stress and brain leukocyte infiltration.
  • Enhances angioneurogenesis.
  • Suppresses calcium influx.
  • Stabilizes neuronal resting membrane potential.
  • Counteracts focal cerebral ischemia.
  • Exhibits analgesic and muscle relaxant properties.
  • Protects neurons from excitotoxic, ischemic, or cytokine-mediated death.
Catalog No. 50-003-90676 Supplier Medchemexpress LLC Supplier No. HY17001100MG
Flupirtine Maleate is an orally active, blood-brain barrier-crossing non-opioid analgesic and neuroprotective agent. It functions as a neuronal potassium channel opener (Kv7 activator), an NMDA receptor antagonist, and a GABA receptor activator. This agent is suitable for research related to focal cerebral ischemia, pain, Alzheimer's disease, or multiple sclerosis.
  • Stabilizes blood-brain barrier integrity.
  • Reduces oxidative stress and brain leukocyte infiltration.
  • Enhances angioneurogenesis.
  • Suppresses calcium influx.
  • Stabilizes neuronal resting membrane potential.
  • Counteracts focal cerebral ischemia.
  • Exhibits analgesic and muscle relaxant properties.
  • Protects neurons from excitotoxic, ischemic, or cytokine-mediated death.