Learn More
Medchemexpress LLC Flupirtine maleate | 75507-68-5 | 100.0% | 420.39 | 50 MG
SDP
Catalog No. 5000391255
Encompass_Preferred
Flupirtine Maleate is an orally active, blood-brain barrier-crossing non-opioid analgesic and neuroprotective agent. It functions as a neuronal potassium channel opener (Kv7 activator), an NMDA receptor antagonist, and a GABA receptor activator. It stabilizes blood-brain-barrier integrity and protects neurons.
  • Stabilizes blood-brain-barrier integrity and reduces oxidative stress
  • Enhances angioneurogenesis and suppresses calcium influx
  • Stabilizes neuronal resting membrane potential
  • Counteracts focal cerebral ischemia
  • Exhibits analgesic and muscle relaxant properties
  • Protects neurons from excitotoxic, ischemic, or cytokine-mediated death
  • Functions as a non-opioid analgesic
  • Used for research on focal cerebral ischemia, pain, Alzheimer's disease, or multiple sclerosis
Catalog No. 50-003-91255 Supplier Medchemexpress LLC Supplier No. HY1700150MG
Flupirtine Maleate is an orally active, blood-brain barrier-crossing non-opioid analgesic and neuroprotective agent. It functions as a neuronal potassium channel opener (Kv7 activator), an NMDA receptor antagonist, and a GABA receptor activator. It stabilizes blood-brain-barrier integrity and protects neurons.
  • Stabilizes blood-brain-barrier integrity and reduces oxidative stress
  • Enhances angioneurogenesis and suppresses calcium influx
  • Stabilizes neuronal resting membrane potential
  • Counteracts focal cerebral ischemia
  • Exhibits analgesic and muscle relaxant properties
  • Protects neurons from excitotoxic, ischemic, or cytokine-mediated death
  • Functions as a non-opioid analgesic
  • Used for research on focal cerebral ischemia, pain, Alzheimer's disease, or multiple sclerosis