Learn More
Apexbio Technology LLC Flupirtine maleate 75507-68-5 100mg
SDP
Catalog No. 5000089964
Encompass_Preferred
Flupirtine maleate (CAS 75507-68-5) is a non-opioid analgesic compound characterized by its modulatory effects on neuronal excitability It acts primarily by opening neuronal potassium channels leading to membrane stabilization and reduced excitatory neurotransmission Additionally flupirtine exerts indirect antagonism of N-methyl-D-aspartate (NMDA) receptors which contributes to its analgesic and muscle relaxant effects Due to this dual mechanism flupirtine maleate is utilized in research focusing on pain modulation neuroprotection and the investigation of NMDA receptor-mediated pathways
Catalog No. 50-000-89964 Supplier Apexbio Technology LLC Supplier No. B1475100
Flupirtine maleate (CAS 75507-68-5) is a non-opioid analgesic compound characterized by its modulatory effects on neuronal excitability It acts primarily by opening neuronal potassium channels leading to membrane stabilization and reduced excitatory neurotransmission Additionally flupirtine exerts indirect antagonism of N-methyl-D-aspartate (NMDA) receptors which contributes to its analgesic and muscle relaxant effects Due to this dual mechanism flupirtine maleate is utilized in research focusing on pain modulation neuroprotection and the investigation of NMDA receptor-mediated pathways