Learn More
Medchemexpress LLC Flupirtine (Maleate) | 75507-68-5 | 99.9% | 420.39 | 1 ML
SDP
Catalog No. 5000391242
Encompass_Preferred
Flupirtine Maleate is an orally active, blood-brain barrier-crossing non-opioid analgesic and neuroprotective agent. It exhibits analgesic and muscle relaxant properties, and protects neurons from excitotoxic, ischemic, or cytokine-mediated death.
  • Neuronal potassium channel opener (Kv7 activator).
  • NMDA receptor antagonist.
  • GABA receptor activator.
  • Stabilizes blood-brain-barrier integrity.
  • Reduces oxidative stress and brain leukocyte infiltration.
  • Enhances angioneurogenesis.
  • Suppresses calcium influx.
  • Stabilizes neuronal resting membrane potential.
  • Counteracts focal cerebral ischemia.
  • Functions as a non-opioid analgesic without antipyretic or antiphlogistic properties.
  • Shows no relevant affinity to opiate receptor.
Catalog No. 50-003-91242 Supplier Medchemexpress LLC Supplier No. HY1700110MM/1ML
Flupirtine Maleate is an orally active, blood-brain barrier-crossing non-opioid analgesic and neuroprotective agent. It exhibits analgesic and muscle relaxant properties, and protects neurons from excitotoxic, ischemic, or cytokine-mediated death.
  • Neuronal potassium channel opener (Kv7 activator).
  • NMDA receptor antagonist.
  • GABA receptor activator.
  • Stabilizes blood-brain-barrier integrity.
  • Reduces oxidative stress and brain leukocyte infiltration.
  • Enhances angioneurogenesis.
  • Suppresses calcium influx.
  • Stabilizes neuronal resting membrane potential.
  • Counteracts focal cerebral ischemia.
  • Functions as a non-opioid analgesic without antipyretic or antiphlogistic properties.
  • Shows no relevant affinity to opiate receptor.