Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flupirtine Maleate Salt 98.0+%, TCI America™
SDP

Catalog No. F0736100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0736100MG 100 mg
F07361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0736100MG Supplier TCI America Supplier No. F0736100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75507-68-5
Molecular Formula C19H21FN4O6
Molecular Weight (g/mol) 420.397
MDL Number MFCD00941415
InChI Key DPYIXBFZUMCMJM-BTJKTKAUSA-N
Synonym flupirtine maleate, katadolon, flupirtine maleate salt, flupirtin-maleat german, unii-0vci53pk4a, flupirtine maleate usan, 0vci53pk4a, ethyl 2-amino-6-p-fluorobenzyl amino-3-pyridinecarbamate maleate 1:1, ethyl-n-2-amino-6-4-fluorophenylmethylamino pyridin-3-yl carbamate maleate
PubChem CID 6435335
IUPAC Name (Z)-but-2-enedioic acid;ethyl N-[2-amino-6-[(4-fluorophenyl)methylamino]pyridin-3-yl]carbamate
SMILES CCOC(=O)NC1=C(N=C(C=C1)NCC2=CC=C(C=C2)F)N.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 100 mg
Formula Weight 420.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Flupirtine Maleate Salt
RTECSNumber EY8555800
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.