Learn More
Cayman Chemical FlupIrtInmaleate 100mg
SDP
Catalog No. 502964909
Encompass_Preferred
An activator of Kv7/KCNQ; induces relaxation of preconstricted pulmonary arteries isolated from wild type and SERT+ mice; decreases mean right ventricular pressure and right ventricular hypertrophy in hypoxia-induced and SERT+ mouse models of pulmonary arterial hypertension at 30 mg/kg per day; increases the paw withdrawal threshold in a rat model of streptozotocin-induced diabetic neuropathy at 10 mg/kg; increases paw withdrawal latency in a rat model of carrageenan-induced paw inflammation when used in combination with morphine; indirectly antagonizes NMDA receptors via its effects on potassium channels
Catalog No. 50-296-4909 Supplier Cayman Chemical Supplier No. 16674100 MG
An activator of Kv7/KCNQ; induces relaxation of preconstricted pulmonary arteries isolated from wild type and SERT+ mice; decreases mean right ventricular pressure and right ventricular hypertrophy in hypoxia-induced and SERT+ mouse models of pulmonary arterial hypertension at 30 mg/kg per day; increases the paw withdrawal threshold in a rat model of streptozotocin-induced diabetic neuropathy at 10 mg/kg; increases paw withdrawal latency in a rat model of carrageenan-induced paw inflammation when used in combination with morphine; indirectly antagonizes NMDA receptors via its effects on potassium channels