Learn More
AdipoGen Flupropanate
SDP
Supplier:  AdipoGen CDXF0086G005
Encompass_Preferred
Chemical. CAS 756-09-2. Formula C3H2F4O2. MW 146.04. Synthetic. Chloro-carbonic-acid herbicide. Compound can be used as analytical reference material.
Catalog No. 50-256-0882