Learn More
Cayman Chemical Fluprostenol-d4 100ug
SDP
Catalog No. 502992051
Encompass_Preferred
An internal standard for the quantification of fluprostenol by GC- or LC-MS
Catalog No. 50-299-2051 Supplier Cayman Chemical Supplier No. 316767100 UG
An internal standard for the quantification of fluprostenol by GC- or LC-MS