Learn More
Cayman Chemical Fluprostenol-d4 500ug
SDP
Catalog No. 502991235
Encompass_Preferred
An internal standard for the quantification of fluprostenol by GC- or LC-MS
Catalog No. 50-299-1235 Supplier Cayman Chemical Supplier No. 316767500 UG
An internal standard for the quantification of fluprostenol by GC- or LC-MS