Learn More
Cayman Chemical Fluprostenol-d4 50ug
SDP
Catalog No. 502988099
Encompass_Preferred
An internal standard for the quantification of fluprostenol by GC- or LC-MS
Catalog No. 50-298-8099 Supplier Cayman Chemical Supplier No. 31676750 UG
An internal standard for the quantification of fluprostenol by GC- or LC-MS