Learn More
Cayman Chemical Fluprostenol Isopropyl STR
SDP
Catalog No. 502895381
Encompass_Preferred
An internal standard for the quantification of fluprostenol isopropyl ester by GC- or LC-MS
Catalog No. 50-289-5381 Supplier Cayman Chemical Supplier No. 9000936100 UG
An internal standard for the quantification of fluprostenol isopropyl ester by GC- or LC-MS