Learn More
Cayman Chemical Fluprostenol Isopropyl STR
SDP
Supplier:  Cayman Chemical 9000936500 UG
Encompass_Preferred
An internal standard for the quantification of fluprostenol isopropyl ester by GC- or LC-MS
Catalog No. 50-289-5207