Learn More
Cayman Chemical Fluprostenol Lacton DIol 100mg
SDP
Catalog No. 502894351
Encompass_Preferred
A G protein-biased peptide agonist of GLP-1R selectively induces cAMP accumulation in HEK293 cells expressing human GLP-1R overBarrestin recruitment in CHO-K1 cells expressing human GLP-1R (EC50s 0.63 and 79 nM respectively) induces insulin secretion in INS-1 832/3 cells (EC50 6.3 nM) reduces food intake and body weight increases plasma insulin levels and decreases plasma glucose HbA1c glucagon and triglyceride levels in diabetic db/db mice from 1.5-6 nmol/kg
Catalog No. 50-289-4351 Supplier Cayman Chemical Supplier No. 70037100 MG
A G protein-biased peptide agonist of GLP-1R selectively induces cAMP accumulation in HEK293 cells expressing human GLP-1R overBarrestin recruitment in CHO-K1 cells expressing human GLP-1R (EC50s 0.63 and 79 nM respectively) induces insulin secretion in INS-1 832/3 cells (EC50 6.3 nM) reduces food intake and body weight increases plasma insulin levels and decreases plasma glucose HbA1c glucagon and triglyceride levels in diabetic db/db mice from 1.5-6 nmol/kg