Learn More
Cayman Chemical Fluprostenol serinol amide
SDP
Supplier:  Cayman Chemical 1000423610 MG
Encompass_Preferred
An analog of PGF2α 2-glyceryl ester with increased stability
Catalog No. 50-289-0247