Learn More
Medchemexpress LLC Flupyrimin | 1689566-03-7 | 98.1% | C13H9ClF3N3O | 10MG
SDP
Catalog No. 5000259691
Encompass_Preferred
Flupyrimin is an antagonist of insect nicotinic acetylcholine receptors (nAChR) that produces insecticidal effects by targeting the insect nervous system. Supplied as a solid analytical standard, it demonstrates high potency in neuronal assays and is intended for research and analytical use.
Catalog No. 50-002-59691 Supplier Medchemexpress LLC Supplier No. HY14529710MG
Flupyrimin is an antagonist of insect nicotinic acetylcholine receptors (nAChR) that produces insecticidal effects by targeting the insect nervous system. Supplied as a solid analytical standard, it demonstrates high potency in neuronal assays and is intended for research and analytical use.