Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flupyrimin | 1689566-03-7 | 98.1% | C13H9ClF3N3O | 50 MG
SDP

Catalog No. 5000354404
Encompass_Preferred

Flupyrimin is a chemical compound functioning as an antagonist for the insect nicotinic acetylcholine receptor (nAChR). It demonstrates insecticidal effects by specifically targeting the insect nervous system.

  • Antagonist for insect nicotinic acetylcholine receptor (nAChR)
  • Exhibits broad-spectrum insecticidal activity
  • Low toxicity to pollinators and rats
  • Molecular weight: 315.68
  • Chemical formula: C13H9ClF3N3O
  • Purity: 98.1%
  • CAS number: 1689566-03-7
  • Appearance: Off-white to light yellow solid

Catalog No. 50-003-54404 Supplier Medchemexpress LLC Supplier No. HY14529750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flupyrimin is a chemical compound functioning as an antagonist for the insect nicotinic acetylcholine receptor (nAChR). It demonstrates insecticidal effects by specifically targeting the insect nervous system.

  • Antagonist for insect nicotinic acetylcholine receptor (nAChR)
  • Exhibits broad-spectrum insecticidal activity
  • Low toxicity to pollinators and rats
  • Molecular weight: 315.68
  • Chemical formula: C13H9ClF3N3O
  • Purity: 98.1%
  • CAS number: 1689566-03-7
  • Appearance: Off-white to light yellow solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.