Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flupyrimin | 1689566-03-7 | MFCD33395052 | 98.1% | 315.68 g/mol | C13H9ClF3N3O | 5 MG
SDP

Catalog No. 5000220973
Encompass_Preferred

Flupyrimin is a small-molecule insecticide research compound that antagonizes insect nicotinic acetylcholine receptors (nAChR), disrupting neuronal signaling in target insects. It is supplied as a solid for laboratory use and is employed in studies of insect neurophysiology and insecticidal activity.

  • CAS number: 1689566-03-7.
  • Molecular formula: C13H9ClF3N3O.
  • Molecular weight: 315.68 g/mol.
  • Typical purity: 98.1% (supplier reported).
  • Solubility: 250 mg/mL in DMSO; multiple in vivo vehicle formulations reported.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).

Catalog No. 50-002-20973 Supplier Medchemexpress LLC Supplier No. HY1452975MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flupyrimin is a small-molecule insecticide research compound that antagonizes insect nicotinic acetylcholine receptors (nAChR), disrupting neuronal signaling in target insects. It is supplied as a solid for laboratory use and is employed in studies of insect neurophysiology and insecticidal activity.

  • CAS number: 1689566-03-7.
  • Molecular formula: C13H9ClF3N3O.
  • Molecular weight: 315.68 g/mol.
  • Typical purity: 98.1% (supplier reported).
  • Solubility: 250 mg/mL in DMSO; multiple in vivo vehicle formulations reported.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.