Learn More
AdipoGen Flupyrsulfuron-methyl sodium
SDP
Supplier:  AdipoGen CDXF0083M010
Encompass_Preferred
Chemical. CAS 144740-54-5. Formula C15H13F3N5NaO7S. MW 487.3. Synthetic. Systemic and slective herbicide. Mode of action is through inhibition of acetolactate synthase ALS acetohydroxyacid synthase AHAS, which causes a blockade of the biosynthesis of amino acids with branched side chains, such as L-Leucin, L-Isoleucin and L-Valin. Compound can be used as analytical reference material.
Catalog No. 50-256-0888