Learn More
Vector Laboratories WFA, WFL, Fluorescein, 2 mg FL-1351-2MG
SDP
Supplier:  Vector Laboratories FL13512
Encompass
Wisteria Floribunda Lectin (WFA, WFL), Fluorescein, 2 mg, appears to preferentially bind carbohydrate structures terminating in N-acetylgalactosamine-linked alpha or beta to the 3 or 6 position of galactose. It has been used to fractionate lymphocyte populations and elicits the production of lymphokines from murine splenocytes. It has sufficient fluorochromes to provide optimum staining characteristics and is essentially free of unconjugated fluorochromes. Solution preserved with sodium azide.
Catalog No. NC1657443