Learn More
Medchemexpress LLC HY-16973 100mg Medchemexpress, Fluralaner CAS:864731-61-3 Purity:>98%
SDP
Catalog No. 502029223
Encompass_Preferred
Medchemexpress, HY-16973 100mg Fluralaner CAS:864731-61-3 Fluralaner (INN) is a systemic insecticide and acaricide Fluralaner through potent blockage of GABA and L-glutamate gated chloride channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9223 Supplier Medchemexpress LLC Supplier No. HY16973100MG
Medchemexpress, HY-16973 100mg Fluralaner CAS:864731-61-3 Fluralaner (INN) is a systemic insecticide and acaricide Fluralaner through potent blockage of GABA and L-glutamate gated chloride channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.