Learn More
Medchemexpress LLC Fluralaner-13C2,15N,d3 | 864731-61-3 | 98.4% | 562.28 | 1 MG
SDP
Catalog No. 5000391224
Encompass_Preferred
Fluralaner-13C2,15N,d3 is the deuterium, 13C-, and 15-labeled Fluralaner. This systemic insecticide and acaricide works through potent blockage of GABA and L-glutamate gated chloride channels.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-91224 Supplier Medchemexpress LLC Supplier No. HY16973S1MG
Fluralaner-13C2,15N,d3 is the deuterium, 13C-, and 15-labeled Fluralaner. This systemic insecticide and acaricide works through potent blockage of GABA and L-glutamate gated chloride channels.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.