Learn More
Medchemexpress LLC Fluralaner (Standard) | 864731-61-3 | 100.0% | 556.29 | 25 MG
SDP
Catalog No. 5000391524
Encompass_Preferred
Fluralaner (Standard) is the analytical standard of Fluralaner. It is intended for research and analytical applications. Fluralaner (INN) is a systemic insecticide and acaricide that works through potent blockage of GABA and L-glutamate gated chloride channels. It is for research use only and not sold to patients.
  • Serves as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-003-91524 Supplier Medchemexpress LLC Supplier No. 25MGHY16973R
Fluralaner (Standard) is the analytical standard of Fluralaner. It is intended for research and analytical applications. Fluralaner (INN) is a systemic insecticide and acaricide that works through potent blockage of GABA and L-glutamate gated chloride channels. It is for research use only and not sold to patients.
  • Serves as a reference standard for assays
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS