Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Flurbiprofen, 98%

Catalog No. AC454660250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454660250 25 g
AC454660010 1 g
AC454660050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC454660250 Supplier Thermo Scientific Chemicals Supplier No. 454660250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Flurbiprofen, CAS # 5104-49-4, is a monocarboxylic acid-based nonsteroidal anti-inflammatory (NSAID).

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Flurbiprofen is composed of a 2-fluoro-[1,1'-biphenyl-4-yl] moiety attached to propionic acid

• Flurbiprofen works by inhibiting prostaglandin-endoperoxide synthase and reducing the production of prostaglandins and pyrogens

Applications

• Flurbiprofen is useful for studying pathways associated with prostaglandin production

• It is suitable for laboratory research on pain, inflammation, ocular pressure, and postoperative recovery

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5104-49-4
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00079303
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
Synonym flurbiprofen, ansaid, froben, antadys, cebutid, flurofen, anside, flurbiprofene, flurbiprofeno, flurbiprofenum
PubChem CID 3394
ChEBI CHEBI:5130
IUPAC Name 2-(3-fluoro-4-phenylphenyl)propanoic acid
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Specifications

Melting Point 110.0°C to 114.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Merck Index 15,4229
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, readily soluble in most polar solvents
Formula Weight 244.27
Percent Purity 98%
Chemical Name or Material Flurbiprofen
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 225-827-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.