Learn More
Medchemexpress LLC Flurbiprofen-d3 10mg | 1185133-81-6 | 10MG
SDP
Catalog No. 5000243271
Encompass_Preferred
Flurbiprofen-d3 is the deuterium labeled Flurbiprofen Flurbiprofen (dl-Flurbiprofen) is a potent orally active nonsteroidal anti-inflammatory agent (NSAIA/NSAID) with antipyretic and analgesic activities Flurbiprofen is commonly used for the research of inflammatory diseases including osteoarthritis and rheumatoid arthritis Flurbiprofen is a non-selective cyclooxygenase (COX) inhibitor that can be used for the research of colorectal cancer[1 [2 [3
Catalog No. 50-002-43271 Supplier Medchemexpress LLC Supplier No. HY10582S10MG
Flurbiprofen-d3 is the deuterium labeled Flurbiprofen Flurbiprofen (dl-Flurbiprofen) is a potent orally active nonsteroidal anti-inflammatory agent (NSAIA/NSAID) with antipyretic and analgesic activities Flurbiprofen is commonly used for the research of inflammatory diseases including osteoarthritis and rheumatoid arthritis Flurbiprofen is a non-selective cyclooxygenase (COX) inhibitor that can be used for the research of colorectal cancer[1 [2 [3