Learn More
Cayman Chemical Flurbiprofen-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1185133-81-6, 5 mg
SDP
Catalog No. 502937004
Encompass_Preferred
An internal standard for the quantification of flurbiprofen by GC- or LC-MS
Catalog No. 50-293-7004 Supplier Cayman Chemical Supplier No. 252775 MG
An internal standard for the quantification of flurbiprofen by GC- or LC-MS