Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flurbiprofen-d3 (dl-flurbiprofen-d3) | 1185133-81-6 | MFCD09840635 | 99.9% | 247.28 | C15H10D3FO2 | 5 MG
SDP

Catalog No. 5000241920
Encompass_Preferred

Flurbiprofen-d3 is the deuterium-labeled analogue of flurbiprofen supplied as a research-grade analytical standard for quantitative assays and mass spectrometry internal standardization. Provided in a small 5 mg quantity, it offers high chemical purity and isotopic enrichment suitable for accurate quantitation in bioanalytical workflows.

  • Chemical formula C15H10D3FO2
  • Molecular weight 247.28
  • Chemical purity 99.9% (LCMS)
  • Isotopic enrichment 98.2%
  • Suitable as an internal standard for mass spectrometry
  • Provided as a solid 5 mg quantity for analytical use

Catalog No. 50-002-41920 Supplier Medchemexpress LLC Supplier No. HY10582S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flurbiprofen-d3 is the deuterium-labeled analogue of flurbiprofen supplied as a research-grade analytical standard for quantitative assays and mass spectrometry internal standardization. Provided in a small 5 mg quantity, it offers high chemical purity and isotopic enrichment suitable for accurate quantitation in bioanalytical workflows.

  • Chemical formula C15H10D3FO2
  • Molecular weight 247.28
  • Chemical purity 99.9% (LCMS)
  • Isotopic enrichment 98.2%
  • Suitable as an internal standard for mass spectrometry
  • Provided as a solid 5 mg quantity for analytical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.