Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flurbiprofen-d5 (dl-flurbiprofen-d5) | 215175-76-1 | 99.8% | 249.29 g·mol⁻¹ | C15H8D5FO2 | 1 MG
SDP

Catalog No. 5000243727
Encompass_Preferred

Flurbiprofen-d5 (dl-Flurbiprofen-d5) is a deuterium-labeled stable isotope analog of flurbiprofen used as an internal standard for quantitative mass spectrometry and other analytical applications.

  • Purity 99.8%.
  • Molecular weight 249.29 g·mol⁻¹.
  • Chemical formula C15H8D5FO2.
  • Isotope labeling d5 (five deuterium atoms).
  • Available in small powder packs: 1 mg, 5 mg, 10 mg, 50 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for use as an analytical internal standard in mass spectrometry quantification.

Catalog No. 50-002-43727 Supplier Medchemexpress LLC Supplier No. HY10582S11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flurbiprofen-d5 (dl-Flurbiprofen-d5) is a deuterium-labeled stable isotope analog of flurbiprofen used as an internal standard for quantitative mass spectrometry and other analytical applications.

  • Purity 99.8%.
  • Molecular weight 249.29 g·mol⁻¹.
  • Chemical formula C15H8D5FO2.
  • Isotope labeling d5 (five deuterium atoms).
  • Available in small powder packs: 1 mg, 5 mg, 10 mg, 50 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for use as an analytical internal standard in mass spectrometry quantification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.