Learn More
Medchemexpress LLC Flurbiprofen impurity 3 | 137045-30-8 | MFCD05664334 | 216.21 | C13H9FO2 | 1 G
SDP
Catalog No. 5000304123
Encompass_Preferred
Flurbiprofen impurity 3 (CAS 137045-30-8) is an identified impurity of the NSAID flurbiprofen used as a drug intermediate and analytical reference in research and development. Intended for analytical characterization, method development, and impurity profiling; consult the certificate of analysis for purity and storage conditions.
  • Identified impurity for analytical reference and method development.
  • Useful for impurity profiling and drug intermediate studies.
  • Chemical formula C13H9FO2; molecular weight 216.21.
  • Provided in small laboratory quantities suitable for R&D use.
  • Certificate of analysis available for purity and storage details.
Catalog No. 50-003-04123 Supplier Medchemexpress LLC Supplier No. HYW0141361G
Flurbiprofen impurity 3 (CAS 137045-30-8) is an identified impurity of the NSAID flurbiprofen used as a drug intermediate and analytical reference in research and development. Intended for analytical characterization, method development, and impurity profiling; consult the certificate of analysis for purity and storage conditions.
  • Identified impurity for analytical reference and method development.
  • Useful for impurity profiling and drug intermediate studies.
  • Chemical formula C13H9FO2; molecular weight 216.21.
  • Provided in small laboratory quantities suitable for R&D use.
  • Certificate of analysis available for purity and storage details.