Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flurbiprofen, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602789 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
5 kg
10 kg
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-789 10 kg Amber Glass Bottle
18-602-786 1 g Amber Glass Bottle
18-602-787 5 g Amber Glass Bottle
18-602-788 25 g Amber Glass Bottle
18-602-790 100 g Amber Glass Bottle
18-602-791 500 g Amber Glass Bottle
18-602-792 1 kg Amber Glass Bottle
18-602-793 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-602-789 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F112810KGBL
Only null left
Add to Cart
Add to Cart
Request a Quote

F1128, 5104-49-4, C15H13FO2

Spectrum™ Chemical Flurbiprofen, USP is a non-steroidal anti-inflammatory drug used to treat pain and inflammation. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5104-49-4
Molecular Formula C15H13FO2
Molecular Weight (g/mol) 244.27
MDL Number MFCD00079303
InChI Key SYTBZMRGLBWNTM-UHFFFAOYNA-N
IUPAC Name 2-{2-fluoro-[1,1'-biphenyl]-4-yl}propanoic acid
SMILES CC(C(O)=O)C1=CC=C(C(F)=C1)C1=CC=CC=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Melting Range 114°-117°C
Packaging Amber Glass Bottle
Quantity 10 kg
UN Number UN2811
Percent Purity 99 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Flurbiprofen
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.