Learn More
Research Products International Corp Fluridon, 250 Milligrams
SDP
Catalog No. 50213316
Encompass_Preferred
Inhibitor of ABA synthesis.
Catalog No. 50-213-316 Supplier Research Products International Corp Supplier No. F200100.25
Add to Cart
Add to Cart
Inhibitor of ABA synthesis.