Learn More
Cayman Chemical Flurandrenolide, Purity: ≥98%, CAS: 1524-88-5, 10 mg
SDP
Catalog No. 502983477
Encompass_Preferred
A topical corticosteroid with anti-inflammatory actions; activates EGFR with an EC50 value of 23 nM
Catalog No. 50-298-3477 Supplier Cayman Chemical Supplier No. 2093310 MG
A topical corticosteroid with anti-inflammatory actions; activates EGFR with an EC50 value of 23 nM