Learn More
Medchemexpress LLC HY-100956 10mg Medchemexpress, Flurofamide CAS:70788-28-2 Purity:>98%
SDP
Catalog No. 501967793
Encompass_Preferred
Medchemexpress, HY-100956 10mg Flurofamide CAS:70788-28-2 Flurofamide is a potent bacterial urease inhibitor with potential clinical utility in the treatment of infection induced urinary stones. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7793 Supplier Medchemexpress LLC Supplier No. HY10095610MG
Add to Cart
Add to Cart
Medchemexpress, HY-100956 10mg Flurofamide CAS:70788-28-2 Flurofamide is a potent bacterial urease inhibitor with potential clinical utility in the treatment of infection induced urinary stones. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.