Learn More
Med Vet International Fluroxin, Enrofloxacin, 136mg, 50ct
Catalog No. 502827783
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Fluroxin™ (enrofloxacin) is a synthetic, chemotherapeutic agent from the class of the quinolone carboxylic acid derivatives. It has antibacterial activity against a broad spectrum of gram-negative and gram-positive bacteria. Enrofloxacin is rapidly absorbed from the digestive tract, penetrating all measured body tissues and fluids.
Catalog No. 50-282-7783 Supplier Med Vet International Supplier No. RXVFLUROX13650
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Fluroxin™ (enrofloxacin) is a synthetic, chemotherapeutic agent from the class of the quinolone carboxylic acid derivatives. It has antibacterial activity against a broad spectrum of gram-negative and gram-positive bacteria. Enrofloxacin is rapidly absorbed from the digestive tract, penetrating all measured body tissues and fluids.