Learn More
Medchemexpress LLC Fluroxypyr | 69377-81-7 | 98.7% | 25 MG
SDP
Catalog No. 5000414062
Encompass_Preferred
Fluroxypyr is an auxin herbicide used to control annual or perennial weeds. It operates by inducing an overdose or excessive endogenous concentrations of auxin, which disrupts the normal auxin homeostasis and its interactions with other hormones within plant tissues.
  • It acts as an auxin herbicide.
  • It is effective against annual weeds.
  • It is effective against perennial weeds.
  • It causes an overdose of auxin in plants.
  • It disrupts auxin homeostasis.
  • It interferes with hormone interactions in plant tissues.
Catalog No. 50-004-14062 Supplier Medchemexpress LLC Supplier No. 25MGHYB1842
Fluroxypyr is an auxin herbicide used to control annual or perennial weeds. It operates by inducing an overdose or excessive endogenous concentrations of auxin, which disrupts the normal auxin homeostasis and its interactions with other hormones within plant tissues.
  • It acts as an auxin herbicide.
  • It is effective against annual weeds.
  • It is effective against perennial weeds.
  • It causes an overdose of auxin in plants.
  • It disrupts auxin homeostasis.
  • It interferes with hormone interactions in plant tissues.